Orders LinkContactUsLinkComponent
Polypropylene bottles, Corning®
Polypropylene bottles, Corning®
Narrow mouth, polypropylene, 500 ml, GL 25
Catalog # CORN1300P-500
Supplier:   Corning
Polypropylene bottles, Corning®
Catalog # CORN1300P-500
Supplier:  Corning
Polypropylene bottles, Corning®
Polypropylene bottles, Corning®
Narrow mouth, polypropylene, 500 ml, GL 25
Price
88,60 €
Log in for availability
1 * 12 items
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
  • Hollow glass stopper style
  • Reusable
  • Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Documents
Specifications
  • Thread:
    GL 25
  • Capacity:
    500 ml
  • Description:
    Narrow mouth, polypropylene
  • Seq:
    02