Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dibromophenol ≥97%

3,5-Dibromophenol ≥97%

Glass bottle for solids, 10 g
Catalog # H60642.09
Irritant
3,5-Dibromophenol ≥97%
Catalog # H60642.09
CAS Number:  626-41-5
3,5-Dibromophenol ≥97%
3,5-Dibromophenol ≥97%

Glass bottle for solids, 10 g

CAS Number:  626-41-5
Price
221,00 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:626-41-5
  • Formula:C₆H₄Br₂O
  • MDL Number:MFCD06411376
  • Molecular Weight:251.91 g/mol
  • Boiling Pt:274 °C (1013 hPa)
  • Melting Pt:79…83 °C
  • Density:2.1 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications