Mop heads, DryMop™
Dry Mop™ handle, metal, 1524 mm
Catalog # MICRDMH-60M
Supplier: MICRONOVA
Jump to:
Product Details & Documents
For dusting and dry mopping large surface areas.
- Low profile design
- Polyester pad head limits particle generation
- Swivel head for easy maneuvering
- Launderable/Autoclavable covers
Specifications
- Description:Dry Mop™ handle, metal
- Length:1524 mm
- Seq:04