Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Buffers
Biological Buffers
Plasmid DNA buffers
Print
Plasmid DNA buffers
Solution 2, lysis buffer, 250 mL
Catalog # PS002
Certificates
SDS
Supplier:
OMEGA BIO-TEK
Plasmid DNA buffers
Catalog # PS002
Supplier:
OMEGA BIO-TEK
CAS Number:
SDS
Certificates
Return to Top
Plasmid DNA buffers
Solution 2, lysis buffer, 250 mL
Specifications
Jump to:
Specifications
CAS Number:
Price
93,20 €
Log in
for availability
1 * 250 mL
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
SDS
sds_VWRCPS002
Specifications
Specifications
Pk:
250 mL
Description:
Solution 2, lysis buffer
Specifications