Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ICP Multi-element Standard 4 elements at 1000 mg/l in 2% HNO3 1 * 500 mL
Catalog # ACSD91C8.1K.2N.L5
Supplier:
CPACHEM
Expand image
ICP Multi-element Standard 4 elements at 1000 mg/l in 2% HNO3 1 * 500 mL
Catalog # ACSD91C8.1K.2N.L5
Supplier:
CPACHEM