Order Entry
Export
Orders LinkContactUsLinkComponent
Multi-element standard solutions for ion chromatography

Multi-element standard solutions for ion chromatography

Multi-element standards for IC (7 components), 100 mg/l, 250 mL
Catalog # ACSD1521.K1.W.L25
Supplier:   CPACHEM
Multi-element standard solutions for ion chromatography
Catalog # ACSD1521.K1.W.L25
Supplier:  CPACHEM
CAS Number:  
Multi-element standard solutions for ion chromatography
Multi-element standard solutions for ion chromatography

Multi-element standards for IC (7 components), 100 mg/l, 250 mL

CAS Number:  
Price
237,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

This standard solution is intended for ion chromatography.

Multi element standards for Br⁻, Cl-, F⁻, NO₂⁻, NO₃⁻, PO₄³⁻, SO₄²⁻ diluted in H₂O.

Specifications

Specifications

  • Concentration:
    100 mg/l
  • Pk:
    250 mL
  • Description:
    Multi-element standards for IC (7 components)

Specifications