Multi-element standard solutions for ion chromatography
Multi-element standards for IC (7 components), 100 mg/l, 100 mL
Catalog # ACSD1521.K1.W.L1
Supplier: CPACHEM
Multi-element standard solutions for ion chromatography
Catalog # ACSD1521.K1.W.L1
Supplier: CPACHEM
CAS Number:
Jump to:
Product Details & Documents
This standard solution is intended for ion chromatography.
Multi element standards for Br⁻, Cl-, F⁻, NO₂⁻, NO₃⁻, PO₄³⁻, SO₄²⁻ diluted in H₂O.
Specifications
Specifications
- Concentration:100 mg/l
- Pk:100 mL
- Description:Multi-element standards for IC (7 components)



