Order Entry
Export
Orders LinkContactUsLinkComponent
Petri dishes

Petri dishes

Without vents, 120 mm, 14 mm, Sterile, 32×10/bag
Catalog # ANIC00.40.18PACI
Supplier:   Anicrin
Petri dishes
Catalog # ANIC00.40.18PACI
Supplier:  Anicrin
Petri dishes
Petri dishes

Without vents, 120 mm, 14 mm, Sterile, 32×10/bag

Price
318,00 €
Log in for availability
1 * 320 items
Specifications
  • Ø:
    120 mm
  • Packed:
    32×10/bag
  • Sterility:
    Sterile
  • Version:
    Without vents
  • Height:
    14 mm
  • Seq:
    12