Petri dishes
With 3 vents, 90 mm, 15 mm, Sterile, 25×20/bag
: Anicrin
PS. Suitable for all microbiology applications.
- Ø:90 mm
- Packed:25×20/bag
- Sterility:Sterile
- Version:With 3 vents
- Height:15 mm
- Seq:06
PS. Suitable for all microbiology applications.