Formalin 10%, Q Path®, buffered
Pot, 5 ml in a 20 ml plastic pot, 102 x 20 ml
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.
Formaldehyde 4% is approximately equal to Formalin 10%.
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Density:1.013 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-001-8
Documents
Certificates
Search by Lot NumberCutsheet VWRQ10.07040WH0002.pdf Cutsheet VWRQ10.300150WH01.pdf Cutsheet VWRQFOR0070AF59001.pdf
Technical Drawing
FOR0070AF59001
Images 360
Specifications
Specification Test Results
- Appearance:Colourless liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
Specifications
- Pack type:Pot
- Pk:102 x 20 ml
- Pk Info:5 ml in a 20 ml plastic pot



