Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Mgcl 25mm can be used for optimization of magnesium ion concentration in pcr reaction. 1 * 1 KIT
Catalog # ABNOD0023-2
Supplier:
Abnova
Expand image
Mgcl 25mm can be used for optimization of magnesium ion concentration in pcr reaction. 1 * 1 KIT
Catalog # ABNOD0023-2
Supplier:
Abnova