ContactUsLinkComponent
Areometer, density, , measuring range:0.700-0.850:0.005g/cm , reference temperature:20°C, length:180mm, without thermometer 1 * 1 items
Catalog # AMARH801280S
Supplier:  Amarell
undefined
Areometer, density, , measuring range:0.700-0.850:0.005g/cm , reference temperature:20°C, length:180mm, without thermometer 1 * 1 items
Catalog # AMARH801280S
Supplier:  Amarell