Digital dry block heaters
Dry Block Heater 3, 3, 330 W, RT*...120 °C, ±0,2 °C, 151×380×73 mm, 1,6 kg
Catalog # 460-1006
Supplier: IKA
Jump to:
Product Details & Documents
Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sensor allows temperature control directly in the sample vessel. It is used with PCR tubes, PCR strips, Greiner tubes, microplates and cuvettes. The block heaters are ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood/urea nitrogen determinations and in situ hybridisations.
- Stepless adjustable temperature from ambient up to 120 °C
- Timer: Countdown, adjustable from 1 min to 99h 59 min
- Counter: Shows heating time
- Highly versatile with a large number of aluminium alloy blocks
- Fixed safety circuit and error code display
- Acoustic signal at end of test
Delivery information: Supplied with PT1000 temperature sensor, but without heating blocks, blocks must be ordered separately.
Caution: * RT = Ambient
Specifications
- Heating capacity:330 W
- No. of blocks:3
- Temperature Range:RT*...120 °C
- Weight:1,6 kg
- Seq:04
- W×D×H:151×380×73 mm
- Temp. stability:±0,2 °C
- Type:Dry Block Heater 3
Accessories
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×11,5 mm Ø holesSingle block, 20×11,5 mm Ø holes460-1009Eppendorf Tubes®, 1,5 ml36,9 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #TypeRangeAvailabilityPrice
- Temperature sensor, coated (SafeCoat), DIN 45322Temperature sensor, coated (SafeCoat), DIN 45322460-1041PT 1000.90−10...+260 °CLog in for availability1 * 1 items - 239,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 80× 6,4 mm Ø stripsSingle block, 80× 6,4 mm Ø strips460-1036PCR tube strips (10×8 strips)15,5 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 30×7,9 mm Ø holesSingle block, 30×7,9 mm Ø holes460-1008PCR tubes, 0,5 ml27,6 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 4×35 mm Ø holesSingle block, 4×35 mm Ø holes460-1025Tubes, 35 mm, with round bottom47,6 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 12×17,5 mm Ø holesSingle block, 12×17,5 mm Ø holes460-1022Tubes, 15 - 16 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Double block
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 30×8,3 mm Ø holesSingle block, 30×8,3 mm Ø holes460-1018Tubes, 6 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 8×23,8 mm Ø holesSingle block, 8×23,8 mm Ø holes460-1032Vials, 23 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 16×13,9 mm Ø holesSingle block, 16×13,9 mm Ø holes460-1020Tubes, 12 - 13 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #AvailabilityPrice
- Support rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 threadSupport rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 thread442-2041Log in for availability1 * 1 items - 88,50 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 12×17,8 mm Ø holesSingle block, 12×17,8 mm Ø holes460-1029Vials, 17 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 5×29 mm Ø holesSingle block, 5×29 mm Ø holes460-1014Conical tubes, 50 ml47,6 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Double blockDouble block460-104096- or 384-well plate (1)13,5 mmLog in for availability1 * 1 items - 214,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, combination: 4/3/2×1,5/15/50 mlSingle block, combination: 4/3/2×1,5/15/50 ml460-1016Centrifuge tubes, 1,5 ml/15 ml/50 ml39,1/44,5/47,6 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×15,8 mm Ø holesSingle block, 20×15,8 mm Ø holes460-1028Vials, 15 mm35 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 6×28,8 mm Ø holesSingle block, 6×28,8 mm Ø holes460-1034Vials, 28 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 8×25,8 mm Ø holesSingle block, 8×25,8 mm Ø holes460-1033Vials, 25 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×13,9 mm Ø holesSingle block, 20×13,9 mm Ø holes460-1021Tubes, 12 - 13 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 24×10,7 mm Ø holesSingle block, 24×10,7 mm Ø holes460-1019Tubes, 10 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×10,9 mm Ø holesSingle block, 20×10,9 mm Ø holes460-1012Corning tubes, 2,0 ml38,1 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×11,5 mm Ø holesSingle block, 20×11,5 mm Ø holes460-1011VWR/Eppendorf Tubes®, 2,0 ml38,1 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 2×6 cuvettes 12,5 mm ØSingle block, 2×6 cuvettes 12,5 mm Ø460-1039Cuvettes25,4 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 8×21 mm Ø holesSingle block, 8×21 mm Ø holes460-1023Tubes, 20 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #AvailabilityPrice
- Bosshead clamp
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 12×19,1 mm Ø holesSingle block, 12×19,1 mm Ø holes460-1026Tubes, 17 - 18 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, combination: 6/10/5×0,5/1,5/2 mlSingle block, combination: 6/10/5×0,5/1,5/2 ml460-1017Microtubes, 0,5 ml/1,5 ml/2,0 ml27,6/39,1/38,1 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 12×19,7 mm Ø holesSingle block, 12×19,7 mm Ø holes460-1030Vials, 19 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×12,7 mm Ø holesSingle block, 20×12,7 mm Ø holes460-1027Vials, 12 mm30 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 12×17,1 mm Ø holesSingle block, 12×17,1 mm Ø holes460-1013Conical tubes, 15 ml44,5 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 6×26,2 mm Ø holesSingle block, 6×26,2 mm Ø holes460-1024Tubes, 25 mm, with round bottom48,4 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 64× 6,4 mm Ø holesSingle block, 64× 6,4 mm Ø holes460-1037PCR tubes (64)20,2 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #AvailabilityPrice
- Holding rod, for fastening ETS-D5 or VT-5 to the support rodHolding rod, for fastening ETS-D5 or VT-5 to the support rod431-2888Log in for availability1 * 1 items - 39,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 9×21,7 mm Ø holesSingle block, 9×21,7 mm Ø holes460-1031Vials, 21 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, combination: 6/5/3×6/12/13/25 mm Ø holesSingle block, combination: 6/5/3×6/12/13/25 mm Ø holes460-1015Test tubes, 6 ml/12 - 13 ml/25 ml48,4/48,4/48,4 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 20×11,1 mm Ø holesSingle block, 20×11,1 mm Ø holes460-1010Microtubes 1,5 ml39,1 mmLog in for availability1 * 1 items - 140,00 €
Product Information
- Catalog #ForDepthAvailabilityPrice
- Single block, 15×16,4 mm Ø holesSingle block, 15×16,4 mm Ø holes460-1035Vials, 16 mm45 mmLog in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #TypeRangeAvailabilityPrice
- Stainless steel temperature sensor, DIN 45322Stainless steel temperature sensor, DIN 45322442-0584PT 1000.60−20...+350 °CLog in for availability1 * 1 items - 169,00 €