Cleanroom labels, matte
Removable cleanroom labels, matte finish, PP, Acrylic, White, 76×51 mm, 1000 labels/roll
Catalog # 817-0305
Supplier: Texwipe
Jump to:
Product Details & Documents
Matte finish cleanroom labels provide excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue.
- Ideal for handwritten applications
Specifications
- Colour:White
- Description:Removable cleanroom labels, matte finish
- Dimensions:76×51 mm
- Material:PP
- Seq:02
- Packed:1000 labels/roll
- Adhesive:Acrylic