Orders LinkContactUsLinkComponent
Cleanroom labels, matte
Cleanroom labels, matte
Removable cleanroom labels, matte finish, PP, Acrylic, White, 102×51 mm, 1000 labels/roll
Catalog # 817-0308
Supplier:   Texwipe
Cleanroom labels, matte
Catalog # 817-0308
Supplier:  Texwipe
Cleanroom labels, matte
Cleanroom labels, matte
Removable cleanroom labels, matte finish, PP, Acrylic, White, 102×51 mm, 1000 labels/roll
Price
840,67 €
Log in for availability
1 * 4 Roll
Product Details & Documents
Matte finish cleanroom labels provide excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue.
  • Ideal for handwritten applications
Specifications
  • Colour:
    White
  • Description:
    Removable cleanroom labels, matte finish
  • Dimensions:
    102×51 mm
  • Material:
    PP
  • Seq:
    03
  • Packed:
    1000 labels/roll
  • Adhesive:
    Acrylic