Order Entry
Export
Orders LinkContactUsLinkComponent
Contrast clips

Contrast clips

PP, 25,5 - 29,5 mm, 36 mm, White
Catalog # WITG3.691.006
Supplier:   witeg Labortechnik
Contrast clips
Catalog # WITG3.691.006
Supplier:  witeg Labortechnik
Contrast clips
Contrast clips

PP, 25,5 - 29,5 mm, 36 mm, White

Price
4,90 €
Log in for availability
1 * 1 items
Product Details & Documents

PP, white/black.

  • For attachment to all measuring flasks
  • With grip deepening
  • Unbreakable, resistant to acids and alkalis
  • For better recognition and adjustment of the liquid level
Specifications
  • Spanning width:
    25,5 - 29,5 mm
  • Colour:
    White
  • Length:
    36 mm
  • Material:
    PP
  • Seq:
    06