Contrast clips
PP, 13,8 - 17,0, 36 mm, White
Catalog # WITG3.691.004
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
PP, white/black.
- For attachment to all measuring flasks
- With grip deepening
- Unbreakable, resistant to acids and alkalis
- For better recognition and adjustment of the liquid level
Specifications
- Spanning width:13,8 - 17,0
- Colour:White
- Length:36 mm
- Material:PP
- Seq:04