Acetone 99.6% ACS
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Acetone 99.6% ACS
Catalog # ACRO423240025
Supplier: Thermo Fisher Scientific
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
ACRO42324
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Isopropyl alcohol:≤0.05 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0006 meq/g
- Aldehyde (HCHO):≤0.002 %
- Methanol:≤0.05 %
- KMnO4-reducing substance:Passes test
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Cat. No.:ACRO423240025
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



