Sodium peroxide ACS
Plastic bottle, 500 g
: di-Sodium peroxide
- :1313-60-6
- :Na₂O₂
- :MFCD00003497
- :77.98 g/mol
- :13,08728
- :460 °C (dec.)
- :2.8 g/cm³
- :Ambient
- :215-209-4
- :1504
- :5.1,I
- Appearance:Yellow Powder or granules (May contain a small quantity of discolored material from the manufacturing process)
- Titration with KMnO4:≥93.0 %
- Chloride (Cl):≤20 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤10 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤20 ppm
- Pack type:Plastic bottle
- Pk:500 g
- Cat. No.:ACRO315765000



