Lead(II) carbonate ACS
Plastic bottle, 100 g
: Lead carbonat
- :598-63-0
- :PbCO₃
- :MFCD00064631
- :267.21 g/mol
- :400 °C
- :6,43 g/cm³ (20 °C)
- :Ambient
- :3077
- :9,III
- Appearance:White Powder
- Chloride (Cl):≤0.002 %
- Nitrate and Nitrite (as NO3):(limit about 0.005%) Passes test
- Cadmium (Cd):≤0.002 %
- Calcium (Ca):≤0.01 %
- Iron (Fe):≤0.005 %
- Potassium (K):≤0.02 %
- Sodium (Na):≤0.05 %
- Zinc (Zn):≤0.003 %
- Insoluble matter:≤0.02 % (in dilute acetic acid)
- Pack type:Plastic bottle
- Pk:100 g



