Order Entry
Export
Orders LinkContactUsLinkComponent
Palmitic acid 98%

Palmitic acid 98%

Plastic bottle, 250 g
Catalog # ACRO129702500
Palmitic acid 98%
Catalog # ACRO129702500
CAS Number:  57-10-3
Palmitic acid 98%
Palmitic acid 98%

Plastic bottle, 250 g

CAS Number:  57-10-3
Price
27,50 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256.43 g/mol
  • Merck Index:13,07064
  • Boiling Pt:351.5 °C (760 mmHg)
  • Melting Pt:59…63 °C
  • Density:0,853 g/cm³ (20 °C)
  • Flash Pt:206 °C
  • Storage Temperature:Ambient
  • EINECS:200-312-9

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder or granules or flakes
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Acid value:
    215 to 233 mg KOH/g
  • Iodine value:
    ≤1 g I2/100 g
  • Saponification value:
    216 to 224 mg KOH/g

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications