Push-in caps for tubes
LDPE, Caps, Ø 22 mm, Natural
Catalog # SEMA7702
Supplier: SEMADENI
Jump to:
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifications
- Colour:Natural
- Description:Caps, Ø 22 mm
- Material:LDPE
- Seq:18