Orders LinkContactUsLinkComponent
HPLC syringes
HPLC syringes
Autosampler syringe, 22, 250 µl, 50 mm, LCQ
Catalog # 555-8224
HPLC syringes
Catalog # 555-8224
HPLC syringes
HPLC syringes
Autosampler syringe, 22, 250 µl, 50 mm, LCQ
Price
238,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
  • Easy and accurate
  • Reproducible manual injections
Applications include industrial chromatography.
Specifications
  • For:
    LCQ
  • Gauge:
    22
  • Volume:
    250 µl
  • Description:
    Autosampler syringe
  • Length:
    50 mm
  • Seq:
    06