Orders LinkContactUsLinkComponent
Vials for environmental analysis, WHEATON®
20,00 ml, 28×60 mm, Clear, Yes, Open top, 24-400, PP, White, PTFE faced silicone
  WHEAW224609
Vials for environmental analysis, WHEATON®
  WHEAW224609
Vials for environmental analysis, WHEATON®
Vials for environmental analysis, WHEATON®
20,00 ml, 28×60 mm, Clear, Yes, Open top, 24-400, PP, White, PTFE faced silicone
354,00 €
1 * 72 items
  • Cap included:
    Yes
  • Cap material:
    PP
  • Cap Size:
    24-400
  • Ø×H:
    28×60 mm
  • Septum:
    PTFE faced silicone
  • Capacity:
    20,00 ml
  • Colour:
    Clear
  • Seq:
    01
  • Cap type:
    Open top
  • Cap colour:
    White