Electrode solutions and reagents for ion selective electrodes
Electrolyte solution 0,1 M, 125 ml, for sensION+ ISE for sodium ion in range 0,023 - 23000 mg/lL, 125 mL
Catalog # HACHLZW9901.99
Supplier: Hach
Electrode solutions and reagents for ion selective electrodes
Catalog # HACHLZW9901.99
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:125 mL
- Description:Electrolyte solution 0,1 M, 125 ml, for sensION+ ISE for sodium ion in range 0,023 - 23000 mg/lL



