ContactUsLinkComponent
Labwash Premium Neutraliz, neck 51 mm, alkaline detergent for washing machines 1 * 20 l
Catalog # 84718.415
Supplier:  VWR Chemicals
undefined
Labwash Premium Neutraliz, neck 51 mm, alkaline detergent for washing machines 1 * 20 l
Catalog # 84718.415
Supplier:  VWR Chemicals
Related Information