Orders LinkContactUsLinkComponent
Microtome blades
Microtome blades
Microtome blades DB80 LS, premium low profile
Catalog # 631-1082
Supplier:   LEICA HISTOLOGY
Microtome blades
Catalog # 631-1082
Supplier:  LEICA HISTOLOGY
Microtome blades
Microtome blades
Microtome blades DB80 LS, premium low profile
Price
139,00 €
Log in for availability
1 * 50 items
Product Details & Documents
With these blades it is easy to section a wide range of tissues. Edge encourages immediate ribboning. Premium edge geometry with 35° angle, so no adjustments are needed for the microtome. The uniform coating plus fine and consistent honing helps eliminating sectioning variation. A durable cutting edge is warranted. Quality of each section block-after-block. Expected results with each blade in the dispenser and between different lots will be achieved. There are no sharp corners. When the blade is positioned in the blade holder assembly of the microtome, the blade is fully covered except for the cutting edge.
  • Sharp edge
  • Durability
  • Consistency
  • User safety
Specifications
  • Description:
    Microtome blades DB80 LS, premium low profile