Orders LinkContactUsLinkComponent
Micro cover glasses
Micro cover glasses
Cover glass 2450-1.0
Catalog # 630-2096
Supplier:   LEICA HISTOLOGY
Micro cover glasses
Catalog # 630-2096
Supplier:  LEICA HISTOLOGY
Micro cover glasses
Cover glass 2450-1.0
Price
110,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
These micro cover glasses feature a fine pre-cleaned borosilicate glass.
  • Uniform thinness and flatness
  • Non corrosive, anti-fogging glass in a lint-free, shock-absorbent lining
  • Strict specification control during the cutting, cleaning, weighing and packing procedures
Packaging: Includes dessicant.
Specifications
  • Description:
    Cover glass 2450-1.0