Micro cover glasses
Cover glass 4570-1.5
Catalog # 630-2098
Supplier: LEICA HISTOLOGY
Jump to:
Product Details & Documents
These micro cover glasses feature a fine pre-cleaned borosilicate glass.
- Uniform thinness and flatness
- Non corrosive, anti-fogging glass in a lint-free, shock-absorbent lining
- Strict specification control during the cutting, cleaning, weighing and packing procedures
Packaging: Includes dessicant.
Specifications
- Description:Cover glass 4570-1.5