Orders LinkContactUsLinkComponent
Accessories for FlashGel™ system
FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
Catalog # LONZ57064
Supplier:   LONZA
Accessories for FlashGel™ system
Catalog # LONZ57064
Supplier:  LONZA
Accessories for FlashGel™ system
Accessories for FlashGel™ system
FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
Price
736,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications
  • Description:
    FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
  • Seq:
    22