Orders LinkContactUsLinkComponent
Nessler cylinders
Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 100 ml, Ø×L: 32×220 mm
Catalog # WITG2.150.100
Supplier:   witeg Labortechnik
Nessler cylinders
Catalog # WITG2.150.100
Supplier:  witeg Labortechnik
Nessler cylinders
Nessler cylinders
Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 100 ml, Ø×L: 32×220 mm
Price
28,20 €
Log in for availability
1 * 1 items
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Specifications
  • Description:
    Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 100 ml, Ø×L: 32×220 mm
  • Seq:
    02
  • Cat. No.:
    WITG2.150.100