Orders LinkContactUsLinkComponent
Shell jacket, GEN-Y Cocona®
Shell jacket, GEN-Y Cocona®
Black, XL, Regular
Catalog # KWIN100963940XL
Supplier:   FRISTADS KANSAS
Shell jacket, GEN-Y Cocona®
Catalog # KWIN100963940XL
Supplier:  FRISTADS KANSAS
Shell jacket, GEN-Y Cocona®
Shell jacket, GEN-Y Cocona®
Black, XL, Regular
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Breathable jacket made of 100% polyester with 3,5-layer Cocona® PU laminate fabric. The fabric is windproof and waterproof.
  • Dot-reinforcement at shoulders, front and lower sleeves
  • Fleece-lined collar, outer and inside storm flap over front zip
  • Double front pockets with logo zip, two inside pockets
  • Raglan sleeves, reflector piping at hem
  • Detachable hood with snap fasteners
  • Adjustable strap with reflector tape on sleeves
Weight: 119 g/m²
Specifications
  • Clothing Size:
    XL
  • Garment fit:
    Regular
  • Colour:
    Black
  • Seq:
    19
  • Cat. No.:
    KWIN100963940XL
  • Sleeve or leg length:
    Long
  • Risk area:
    Rain (waterproof)
  • Model:
    Gen-Y
  • Garment application:
    Outdoor
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Fabric:
    Polyester
  • Weight:
    119 g/m²