Reusable earplugs, Comets®
Detectable earplugs, in pocket case, Blue, L
Catalog # 111-1400
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
These reusable earplugs are metal detectable and blue coloured making them perfect for use in the food industry. They provide all-day comfort and superb attenuation.
- Detectable ring on stem and attached metalised cord detectable by automated equipment
- FlexiFirm™ core and soft TPE flanges ensure easy insertion into the ear canal and a superior fit
- Detachable cord made of soft braided fabric
SNR: 28 dB
Certifications: EN 352-2
Specifications
- Colour:Blue
- Description:Detectable earplugs, in pocket case
- Seq:02
- Size:L