Orders LinkContactUsLinkComponent
Goggles, DuraMaxx™
Goggles, DuraMaxx™
Anti-scratch, anti-fog; with fabric headband, Clear PC, Blue/grey
Catalog # 111-1435
Supplier:   HONEYWELL SAFETY
Goggles, DuraMaxx™
Catalog # 111-1435
Supplier:  HONEYWELL SAFETY
Goggles, DuraMaxx™
Goggles, DuraMaxx™
Anti-scratch, anti-fog; with fabric headband, Clear PC, Blue/grey
Price
25,30 €
Log in for availability
1 * 1 items
Product Details & Documents
These low profile goggles with toric lens provide superior optics and unobstructed vision, while the soft, elastomer bodies flex and conform to the face for a comfortable, gap-free fit.
  • PP/TPE frame and large adjustable headband
  • Indirect venting system minimises fogging to keep lenses clear
  • Dura-streme™ lens coating provides anti-scratch coating on the outside and anti-fog coating on the inside
The OTG goggles fit over most prescription spectacles comfortably. Clear lens model is adapted for indoor use, grey model for outdoor use.
Weight: 100 g
Certifications: EN 166
Specifications
  • Type:
    Anti-scratch, anti-fog; with fabric headband
  • Colour:
    Blue/grey
  • Seq:
    1
  • Lenses:
    Clear PC