Order Entry
Export
Orders LinkContactUsLinkComponent
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%

(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%

Glass bottle for solids, 1 g
Catalog # H35096.03
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%
Catalog # H35096.03
CAS Number:  7326-19-4
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%

Glass bottle for solids, 1 g

CAS Number:  7326-19-4
Price
28,20 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid
  • CAS Number:7326-19-4
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00078062
  • Molecular Weight:166.1739 g/mol
  • EINECS:230-803-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications