Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Hydroxyindole ≥98%

Glass bottle for solids, 1 g
Catalog # L14352.03
Irritant
4-Hydroxyindole ≥98%
Catalog # L14352.03
CAS Number:  2380-94-1
4-Hydroxyindole ≥98%
4-Hydroxyindole ≥98%

Glass bottle for solids, 1 g

CAS Number:  2380-94-1
Price
30,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 2380-94-1 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 97-99° Beilstein Reference: 114905

Synonyms:  Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
  • CAS Number:2380-94-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00005667
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:100 °C (0.01 mmHg)
  • Melting Pt:97…99 °C
  • Storage Temperature:Ambient
  • EINECS:219-177-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications