Funnels, powder, Nalgene®
100 mm, 23 mm, PP, 34 mm, Powder funnels, 243 ml, 105 mm hieght
PPCO, translucent.
- Very good chemical resistance
- Outer ribs prevent air from being trapped
- Autoclavable
For transferring powdery materials. Parallel stem prevents blockages.
This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
- Funnel O-Ø:100 mm
- Description:Powder funnels, 243 ml, 105 mm hieght
- Material:PP
- Seq:03
- Stem O-Ø:23 mm
- Stem length:34 mm
- Cat. No.:221-0092
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



