Orders LinkContactUsLinkComponent
Dropping bottles, glass
Dropping bottles, glass
10 ml, DIN 18
Catalog # HECH41314010
Dropping bottles, glass
Catalog # HECH41314010
Dropping bottles, glass
Dropping bottles, glass
10 ml, DIN 18
Price
56,80 €
Log in for availability
1 * 198 items
Product Details & Documents
Made of amber glass.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances
Dropping bottles without cap, with thread DIN 18.
Ordering information: Pipette kit and screw cap must be ordered seperately.
Specifications
  • Thread:
    DIN 18
  • Capacity:
    10 ml
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, red, for 5 ml dropping bottles
    Pipette kit, red, for 5 ml dropping bottles
    HECH41315006
    Log in for availability
    1 * 100 items - 55,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, blue, for 20 ml dropping bottles
    Pipette kit, blue, for 20 ml dropping bottles
    HECH41315020
    Log in for availability
    1 * 100 items - 53,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, blue, for 30 ml dropping bottles
    Pipette kit, blue, for 30 ml dropping bottles
    HECH41315030
    Log in for availability
    1 * 100 items - 55,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, red, for 30 ml dropping bottles
    Pipette kit, red, for 30 ml dropping bottles
    HECH41315031
    Log in for availability
    1 * 100 items - 55,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, red, for 100 ml dropping bottles
    Pipette kit, red, for 100 ml dropping bottles
    HECH41315061
    Log in for availability
    1 * 100 items - 56,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, red, for 20 ml dropping bottles
    Pipette kit, red, for 20 ml dropping bottles
    HECH41315021
    Log in for availability
    1 * 100 items - 53,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, red, for 10 ml dropping bottles
    Pipette kit, red, for 10 ml dropping bottles
    HECH41315011
    Log in for availability
    1 * 100 items - 53,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, red, for 50 ml dropping bottles
    Pipette kit, red, for 50 ml dropping bottles
    HECH41315051
    Log in for availability
    1 * 100 items - 54,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette kit, blue, for 50 ml dropping bottles
    Pipette kit, blue, for 50 ml dropping bottles
    HECH41315650
    Log in for availability
    1 * 100 items - 39,00 €