Orders LinkContactUsLinkComponent
FT-IR Spectrometer, Spectrum Two™
Infrared spectrometer, Spectrum Two™, 8,300 - 350 cm-1
  PERKL160000A
FT-IR Spectrometer, Spectrum Two™
  PERKL160000A
FT-IR Spectrometer, Spectrum Two™
Infrared spectrometer, Spectrum Two™, 8,300 - 350 cm-1
30 970,00 €
1 * 1 items
Spectrum Two is a DynaScan interferometer with KBr optics delivers 9,300:1 peak-peak signal-to-noise for five seconds.
Product features include OpticsGuard™ technology, Atmospheric Vapor Compensation™ (AVC).
High performance detector, a standard sample compartment with sample slide mount and the comprehensive Spectrum 10™ and Spectrum 10™ upgrade software interfaces.
L16000F includes the temperature stabilised deuterated triglycine sulphate (DTGS) detector, automatic performance verification (APV) comprising of an integrated fully automatic filter wheel containing traceable reference materials and software allowing abscissa, ordinate, noise, ASTM and USP tests to be performed.
Absolute virtual instrument (AVI) to allow the production of standard data referenced to fundamental band positions is also included using a manual methane reference cell. Spectrum 10™ software suite is also included.
 : Please contact your local Avantor office for specific applications including environmental, phamaceutical, polymer analysis and the corresponding specifications.
 : Supplied with mains cable, Ethernet and USB cables, electronic user’s and maintenance guide on CD, certificate of conformity and fuses. One year warranty and installation guide (additional on-site installation and warranties optional).
  • Model:
    Spectrum Two™
  • Range:
    8,300 - 350 cm-1
  • Description:
    Infrared spectrometer
  • Seq:
    01