Accessories
Retrofittable magnetic spray arms for spray arm for retrofitting upper baskets without magnetic spray arm, for use in O 167, O 173, O 188, O 190, PG 8535/36
Catalog # 141-0571
Supplier: MIELE
Jump to:
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifications
- Description:Retrofittable magnetic spray arms for spray arm for retrofitting upper baskets without magnetic spray arm, for use in O 167, O 173, O 188, O 190
- Seq:29
- For:PG 8535/36