Orders LinkContactUsLinkComponent
Accessories
Accessories
Conductivity meter retrofit kit, PG 8536
Catalog # 141-0580
Supplier:   MIELE
Accessories
Catalog # 141-0580
Supplier:  MIELE
Accessories
Accessories
Conductivity meter retrofit kit, PG 8536
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    Conductivity meter retrofit kit
  • Seq:
    08
  • For:
    PG 8536