Orders LinkContactUsLinkComponent
Caps for sample tubes, tamper evident
Caps for sample tubes, tamper evident
Screw cap, with lip seal for tamper evident tubes, 7 ml, White
Catalog # SIMPT552-7WTP
Supplier:   Simport Scientific
Caps for sample tubes, tamper evident
Catalog # SIMPT552-7WTP
Supplier:  Simport Scientific
Caps for sample tubes, tamper evident
Caps for sample tubes, tamper evident
Screw cap, with lip seal for tamper evident tubes, 7 ml, White
Price
101,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • For tubes:
    7 ml
  • Colour:
    White
  • Description:
    Screw cap, with lip seal for tamper evident tubes
  • Seq:
    02