Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Coenzyme Q-10, 98% 1 * 25 g
Catalog # J65137.14
Supplier:
Thermo Fisher Scientific
SDS
Certificates
Expand image
Coenzyme Q-10, 98% 1 * 25 g
Catalog # J65137.14
Supplier:
Thermo Fisher Scientific
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ65137
Application Notes
Organic Building Blocks and Intermediates
Documentation