Bisacrylamide 2% in aqueous solution
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Bisacrylamide 2% in aqueous solution
Catalog # J63265.K2
Supplier: Thermo Fisher Scientific
CAS Number: 110-26-9
Jump to:
Product Details & Documents
Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS Number:110-26-9
- Formula:(H₂C=CHCONH)₂CH₂
- MDL Number:MFCD00008625
- Molecular Weight:154.17 g/mol
- Storage Temperature:Refrigerator
- EINECS:203-750-9
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



