Order Entry
Export
Orders LinkContactUsLinkComponent
 

20-Hydroxyecdysone

50 mg
Catalog # J63091.MF
20-Hydroxyecdysone
Catalog # J63091.MF
CAS Number:  5289-74-7
20-Hydroxyecdysone
20-Hydroxyecdysone

50 mg

CAS Number:  5289-74-7
Price
810,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

CAS No.: 5289-74-7 Molecular Formula: C27H44O7 Formula Weight: 480.64 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 242-244° Solubility: Soluble in alcohol Beilstein Reference: 1917578 MDL No.: MFCD00036740

Applications: An insect steroid hormone that induces apoptosis

Synonyms:  β-Ecdysone
  • CAS Number:5289-74-7
  • Formula:C₂₇H₄₄O₇
  • MDL Number:MFCD00036740
  • Molecular Weight:480.642 g/mol
  • Merck Index:14,3491
  • Melting Pt:242…244 °C
Specifications

Specifications

  • Pk:
    50 mg

Specifications