Order Entry
Export
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 150,000

Dextran, M.W. ~ 150,000

Glass bottle for solids, 100 g
Catalog # J63789.22
Dextran, M.W. ~ 150,000
Catalog # J63789.22
CAS Number:  9004-54-0
Dextran, M.W. ~ 150,000
Dextran, M.W. ~ 150,000

Glass bottle for solids, 100 g

CAS Number:  9004-54-0
Price
156,00 €
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 9004-54-0 Molecular Formula: (C6H16O5)n Physical Form: Powder Appearance: White Merck Reference: 14,2948 MDL No.: MFCD00130935

  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • Merck Index:14,2948
  • Storage Temperature:Ambient
  • EINECS:232-677-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications