Hygromycin B
Glass bottle for solids, 250 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS Number:31282-04-9
- Formula:C₂₀H₃₇N₃O₁₃
- MDL Number:MFCD06795479
- Molecular Weight:527.5 g/mol
- Merck Index:14,4852
- Storage Temperature:Refrigerator
- EINECS:250-545-5
- UN:3462
- ADR:6.1,I
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product Literature
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



