RBC lysis buffer for mouse
RBC lysis buffer for mouse, 500 mL
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Applications: For lysis of mouse red blood cells
- Pk:500 mL
- Description:RBC lysis buffer for mouse
- Cat. No.:J62150.AP
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- Appearance:Colourless



