Order Entry
Export
Orders LinkContactUsLinkComponent
RBC lysis buffer for mouse

RBC lysis buffer for mouse

RBC lysis buffer for mouse, 250 mL
Catalog # J62150.AK
RBC lysis buffer for mouse
Catalog # J62150.AK
CAS Number:  
RBC lysis buffer for mouse
RBC lysis buffer for mouse

RBC lysis buffer for mouse, 250 mL

CAS Number:  
Price
72,10 €
Log in for availability
1 * 250 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Applications: For lysis of mouse red blood cells

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    RBC lysis buffer for mouse
  • Cat. No.:
    J62150.AK
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications