Buffers, TRIS, 0,5 M solutions
TRIS buffer solution, 0,5 mol/L, 8,5, Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 77-86-1
MDL No.: MFCD00004679
Documents
Specifications
Specifications
- Concentration:0,5 mol/L
- Pack type:Glass bottle
- Pk:1 L
- Description:TRIS buffer solution
- pH value:8,5
- Cat. No.:J62131.K2
- Physical form:Liquid
- MDL No.:MFCD00004679
- CAS No.:77-86-1



